Order Entry
United States
ContactUsLinkComponent
CT-2000 Series Constant Temperature Bath, 120 V, Cannon Instrument Company
CT-2000 Series Constant Temperature Bath, 120 V, Cannon Instrument Company
Catalog # 47735-074
Supplier:  Cannon
Small Business Enterprise
undefined
CT-2000 Series Constant Temperature Bath, 120 V, Cannon Instrument Company
Catalog # 47735-074
Supplier:  Cannon
Supplier Number:  9726-A30
Small Business Enterprise

Specifications

  • Description:
    CT-2000 Constant Temperature Bath
  • Electrical:
    120V, 115-120VAC, 50/60Hz
  • Cat. No.:
    47735-074
  • Supplier no.:
    9726-A30

Specifications

About this item

Convenient keypad operation and ASTM reliability at temperatures to 150°C make the CT-2000 the bath of choice for many users

Microprocessor-controlled temperature regulation, on-screen temperature monitoring, and serial networking options make this digital bath an attractive and fully functional addition to the modern laboratory.