ContactUsLinkComponent
CT-518 Series Constant Temperature Bath, 115 V, Cannon Instrument Company
CT-518 Series Constant Temperature Bath, 115 V, Cannon Instrument Company
  47735-062
 :  Avantor
Small Business Enterprise
undefined
CT-518 Series Constant Temperature Bath, 115 V, Cannon Instrument Company
  47735-062
 :  Avantor
Small Business Enterprise
  • Description:
    115VAC
  • Cat. No.:
    47735-062
  • Supplier no.:
    9726-A17
Specifically designed for precise viscosity determination with glass capillary viscometers the CT-500 bath offers superior temperature control to 100°C
The CT-500 offers a 45.7 cm (18") bath jar that can accommodate most viscometers.