Order Entry
United States
ContactUsLinkComponent
CT-500 Series Constant Temperature Bath, 230 V, Cannon Instrument Company
CT-500 Series Constant Temperature Bath, 230 V, Cannon Instrument Company
Catalog # 47735-060
Supplier:  Cannon
Small Business Enterprise
undefined
CT-500 Series Constant Temperature Bath, 230 V, Cannon Instrument Company
Catalog # 47735-060
Supplier:  Cannon
Supplier Number:  9726-A15
Small Business Enterprise

Specifications

  • Description:
    230VAC
  • Cat. No.:
    47735-060
  • Supplier no.:
    9726-A15

Specifications

About this item

Specifically designed for precise viscosity determination with glass capillary viscometers the CT-500 bath offers superior temperature control to 100°C

  • 240 V model, not for use in USA

The CT-500 offers a 30.5 cm (12") bath jar that can accommodate most viscometers.

Caution: 230 V model, not for use in USA. Corresponding model with voltage suitable for use in US (47735-058) is also available, please see separate product page.