Human Recombinant IFN alpha 2B (from E. coli)
Supplier: Avantor
Certificates
About this item
>98% Pure active Recombinant human IFN-alpha 2b.
The specific activity as determined in a viral resistance assay using bovine kidney MDBK cells was found to be 2.6 x 10⁸ units/mg
IFN-α 2b is a lymphoid factor that possesses potent anti-viral activity. Human IFN-α 2b is a 19.3 kDa protein containing 165 amino acid residues.
Specifications
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Endotoxin Content:<0.1 ng/μg
- Gene ID:3440
- Reconstitution Instructions:Reconstitute in H₂O to a concentration of 1 mg/ml.
- Protein Synonyms:MGC125764|Interferon alpha 2b|INFA2|MGC125765|IFNA
- Protein/Peptide Name:IFN alpha 2B
- Purity:>98%
- Molecular Weight:19.3 kDa
- Formulation:Sterile filtered and lyophilized from 5 mM sodium phosphate, pH 7.4.
Specifications
Frequently Bought Together