BS/IP/MSL Miniature Suspended Level Viscometer, Cannon Instrument
Supplier: Cannon
Certificates
About this item
The BS/IP/MSL Miniature Suspended Level Viscometer measures kinematic viscosity of transparent Newtonian liquids according to ASTM D445 and ISO 3104
Viscometer is calibrated. Minimum sample volume 4 mL. Minimum bath depth of 12".
Frequently Bought Together