Orders LinkContactUsLinkComponent
BS/IP/SL(S) Suspended Level Viscometer, Cannon Instrument
BS/IP/SL(S) Suspended Level Viscometer, Cannon Instrument
Supplier:   Cannon
BS/IP/SL(S) Suspended Level Viscometer, Cannon Instrument
Supplier:  Cannon
Certificates
BS/IP/SL(S) Suspended Level Viscometer, Cannon Instrument
BS/IP/SL(S) Suspended Level Viscometer, Cannon Instrument
9 Options Available Below
Product Details & Documents
The BS/IP/SL(S) - Suspended Level Viscometer measures kinematic viscosity of transparent Newtonian liquids according to ASTM D445 and ISO 3104
Viscometer is calibrated. Minimum sample volume 10 mL. Minimum bath depth of 9".
Product Family Options
Product Information
  • Size
    Range
    Approx. constant
    Availability
    Price
  • 10791-574
    Calibrated
    Calibrated
    1
    1.05 minimum cSt
    0.0008 cSt/s
    Log in for availability
    Log in for availability
    Each
    Each - $545.67
    $545.67
    Each
       $545.67
    Minimum order quantity is 1
    Supplier Number: 9724-F50
  • 10791-576
    Calibrated
    Calibrated
    2
    2.1 - 3 cSt
    0.003 cSt/s
    Log in for availability
    Log in for availability
    Each
    Each - $545.67
    $545.67
    Each
       $545.67
    Minimum order quantity is 1
    Supplier Number: 9724-F53
  • 10791-578
    Calibrated
    Calibrated
    3
    3.8 - 10 cSt
    0.01 cSt/s
    Log in for availability
    Log in for availability
    Each
    Each - $545.67
    $545.67
    Each
       $545.67
    Minimum order quantity is 1
    Supplier Number: 9724-F56
  • 10791-290
    Calibrated
    Calibrated
    4
    6 - 30 cSt
    0.03 cSt/s
    Log in for availability
    Log in for availability
    Each
    Each - $545.67
    $545.67
    Each
       $545.67
    Minimum order quantity is 1
    Supplier Number: 9724-F59
  • 10791-292
    Calibrated
    Calibrated
    5
    20 - 100 cSt
    0.1 cSt/s
    Log in for availability
    Log in for availability
    Each
    Each - $545.67
    $545.67
    Each
       $545.67
    Minimum order quantity is 1
    Supplier Number: 9724-F62
  • 10791-294
    Calibrated
    Calibrated
    6
    60 - 300 cSt
    0.3 cSt/s
    Log in for availability
    Log in for availability
    Each
    Each - $545.67
    $545.67
    Each
       $545.67
    Minimum order quantity is 1
    Supplier Number: 9724-F65
  • 10791-296
    Calibrated
    Calibrated
    7
    200 - 1000 cSt
    1 cSt/s
    Log in for availability
    Log in for availability
    Each
    Each - $545.67
    $545.67
    Each
       $545.67
    Minimum order quantity is 1
    Supplier Number: 9724-F68
  • 10791-298
    Calibrated
    Calibrated
    8
    600 - 3000 cSt
    3 cSt/s
    Log in for availability
    Log in for availability
    Each
    Each - $545.67
    $545.67
    Each
       $545.67
    Minimum order quantity is 1
    Supplier Number: 9724-F71
  • 10791-300
    Calibrated
    Calibrated
    9
    2000 - 10000 cSt
    10 cSt/s
    Log in for availability
    Log in for availability
    Each
    Each - $545.67
    $545.67
    Each
       $545.67
    Minimum order quantity is 1
    Supplier Number: 9724-F74