Order Entry
United States
ContactUsLinkComponent
Beta-Lactamase Activity Colorimetric Assay Kit, BioVision
Beta-Lactamase Activity Colorimetric Assay Kit, BioVision
  10191-092
 :  Avantor
 :  
Beta-Lactamase Activity Colorimetric Assay Kit, BioVision
  10191-092
 :  Avantor
 :  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

 

  • Size:
    100 assays
  • Species Reactivity:
    Mammalian samples
  • Shelf Life:
    12 Months
  • Sample Size:
    Serum, urine, saliva from mammals infected with βL-secreting bacteria, Food (e.g. milk) and Fermentation media, bacterial cultures, etc.
  • Storage Temperature:
    -20°C
  • Cat. No.:
    10191-092
  • Supplier no.:
    K803-100

 

 

Detection method: Absorbance (490 nm)
Application:
- Measurement of β-Lactamase activity in various biological samples
- Analysis of β-Lactamase activity in pathological conditions

Beta-Lactamases (βLs), are a large family of hydrolases comprising more than 850 identified members expressed in Gram-positive and Gram-negative bacteria. βLs can be classified according to their substrate or inhibitor specificity. These enzymes are capable of hydrolyzing four atom rings known as β-lactams. Antibiotics containing β-lactam rings (i.e. penicillin, cephalosporin, monobactam, carbapenem) are highly susceptible to be hydrolyzed via enzymatic activity, which deactivates their antibiotic potency. βLs have become a significant clinical threat due to the alarming number of cases of bacterial strains showing β-lactam antibiotic resistance. BioVision’s Beta-Lactamase Activity Assay Kit offers a simple and sensitive assay that can detect and quantify the enzymatic activity of these hydrolases. The assay is based on the hydrolysis of Nitrocefin, a chromogenic cephalosporin, that results in the generation of a colored product (OD 490 nm), which is directly proportional to the amount of βL activity. The assay can detect enzymatic activity as low as 0.06 mU in a variety of biological samples.