BS/U/M Miniature Viscometer, Cannon Instrument
Supplier: Cannon
Certificates
About this item
The BS/U/M Miniature Viscometer measures kinematic viscosity of transparent Newtonian liquids according to ASTM D445 and ISO 3104
Viscometer is calibrated. Minimum sample volume 2 mL. Minimum bath depth of 9".
Frequently Bought Together