To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Size:200 assays
- Species Reactivity:Mammalian samples
- Shelf Life:12 Months
- Sample Size:Cell and tissue culture homogenates (eukaryotic cells), serum, plasma and high protein samples
- Storage Temperature:RT
- Cat. No.:10187-074
- Supplier no.:K808-200
Specifications
About this item
Simple Kit to deproteinize samples
Species reactivity- Mammalian
Application- Unique tool to prepare samples for the analysis of a variety of small molecules, proteins are precipitated, excess PCA is removed, samples are neutralized.
The presence of protein and various enzyme activities frequently interferes with the analysis of small molecules in biological samples. Many bioassays require removal of proteins from samples prior to analysis. Among the deproteinization protocols developed over the last half century, perchloric acid (PCA) precipitation has been extensively used in many different sample preparation procedures, since not only does it remove most of the protein present but it also functions to stabilize many of the small molecule analytes. PCA deproteinization methods have been successfully used in the preparation samples prior to quantitation of an array of small molecules, including glycogen, ATP, cAMP, glutathione, antioxidants, etc.The newly designed Deproteinizing Sample Preparation Kit utilizes a PCA precipitation method, providing a unique tool to prepare samples for the analysis of a variety of small molecules. Using the kit, proteins are precipitated, excess PCA is removed, samples are neutralized. Samples prepared using this kit can be directly used in a wide variety of bioassays. The method is easy, convenient, and can be used for the preparation of a large number of samples.