Order Entry
United States
ContactUsLinkComponent
Glutamate Dehydrogenase Activity Colorimetric Assay Kit, BioVision
Glutamate Dehydrogenase Activity Colorimetric Assay Kit, BioVision
Catalog # 10186-994
Supplier:  Avantor
CAS Number:  
undefined
Glutamate Dehydrogenase Activity Colorimetric Assay Kit, BioVision
Catalog # 10186-994
Supplier:  Avantor
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Size:
    100 assays
  • Environmentally Preferable:
  • Species Reactivity:
    Mammalian samples
  • Shelf Life:
    12 Months
  • Sample Size:
    Cell and tissue culture supernatants, urine, plasma and serum, as well as many other biological fluids etc.
  • Storage Temperature:
    -20°C
  • Cat. No.:
    10186-994
  • Supplier no.:
    K729-100

Specifications

About this item

Detection method- Absorbance (450 nm)
Species reactivity- Mammalian
Application- This assay detects GDH activity as low as 0.01mU in serum or tissue and cell extracts.

Glutamate dehydrogenase (GDH) is an enzyme that converts glutamate to α-Ketoglutarate, and vice versa. It represents a key link between catabolic and metabolic pathways, and is therefore ubiquitous in eukaryotes. BioVision’s GDH Assay Kit provides a convenient tool for sensitive detection of GDH in a variety of samples. GDH in sample will consume glutamate as a specific substrate and generate NADH stoichiometrically, resulting in a proportional color development. The GDH activity is easily quantified colorimetrically (λ = 450 nm). This assay detects GDH activity as low as 0.01mU in serum or tissue and cell extracts.