To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:histocompatibility 2, K1, K region
- Antigen Symbol:H2-K1
- Clonality:Monoclonal
- Clone:34-1-2S
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Mouse
- Size:0.1 mg
- Environmentally Preferable:
- Cross Adsorption:No
- Form:liquid
- Gene ID:P01901
- Antigen Synonyms:H2-K1
- Storage Buffer:Phosphate-buffered aqueous solution, ≤0.09% Sodium azide, may contain carrier protein/stabilizer, ph7.2.
- Storage Temperature:The product should be stored undiluted at 4 °C and should be protected from prolonged exposure to light. Do not freeze.
- Shipping Temperature:4 °C
- Purification:The monoclonal antibody was purified utilizing affinity chromatography and unreacted dye was removed from the product.
- Cat. No.:10765-866
- Supplier no.:77-187
Specifications
About this item
The 34-1-2S monoclonal antibody specifically reacts with the mouse H-2k/H2D MHC class I alloantigens, which are involved in antigen presentation to T cells. The 34-1-2S antibody cross-reacts with the b, p, q, r, and s haplotypes.
Type: Primary
Antigen: H2-K1
Clonality: Monoclonal
Clone: 34-1-2S
Conjugation: FITC
Epitope:
Host: Mouse
Isotype: Mouse IgG2a, kappa
Reactivity: Mouse