Specifications
- Antibody Type:Primary
- Antigen Name:tripartite motif containing 24
- Antigen Symbol:TRIM24
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:0.02 mg
- Cross Adsorption:No
- Form:Liquid
- Gene ID:O15164
- Antigen Synonyms:TRIM24
- Storage Buffer:TRIM24 Antibody is supplied in PBS containing 0.02% sodium azide.
- Molecular Weight:Predicted: 112, 116 Observed: 108, 1 of 2 isoforms
- Storage Temperature:TRIM24 antibody can be stored at 4 °C for three months and –20 °C, stable for up to one year.
- Concentration:1 mg/mL
- Shipping Temperature:4 °C
- Immunogen:TRIM24 antibody was raised against a 19 amino acid peptide near the center of human TRIM24. The immunogen is located within amino acids 530 - 580 of TRIM24.
- Tested Applications:Immunocytochemistry
- Purification:TRIM24 Antibody is affinity chromatography purified via peptide column.
- Cat. No.:10751-988
- Supplier no.:7383_S
Specifications
About this item
TRIM24 Antibody: TRIM24 (TIF1), a member of the tripartite motif (TRIM) family, plays a role in the regulation of cell proliferation and apoptosis. TRIM24 localizes to nuclear bodies and is thought to associate with chromatin and heterochromatin-associated factors. The TRIM motif includes three zinc-binding domains (RING, B-box type 1 and B-box type 2) and a coiled-coil region. TIF1 mediates transcriptional events by interactions with the AF2 region of several nuclear receptors, such as the estrogen, retinoic acid and vitamin D3 receptors. Defects in TRIM24 are a cause of thyroid papillary carcinoma (TPC).
Type: Primary
Antigen: TRIM24
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat