Specifications
- Antibody Type:Primary
- Antigen Name:zinc finger and BTB domain containing 5
- Antigen Symbol:ZBTB5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:0.02 mg
- Cross Adsorption:No
- Form:Liquid
- Gene ID:O15062
- Antigen Synonyms:ZBTB5
- Storage Buffer:ZBTB5 Antibody is supplied in PBS containing 0.02% sodium azide.
- Storage Temperature:ZBTB5 antibody can be stored at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:1 mg/mL
- Shipping Temperature:4 °C
- Immunogen:ZBTB5 antibody was raised against a 16 amino acid synthetic peptide near the carboxy terminus of human ZBTB6. The immunogen is located within the last 50 amino acids of ZBTB5.
- Purification:ZBTB5 Antibody is affinity chromatography purified via peptide column.
- Cat. No.:10751-196
- Supplier no.:6123_S
Specifications
About this item
ZBTB5 Antibody: The ZBTB family of proteins is comprised of diverse zinc finger proteins that also contain a BTB (BR-C, ttk and bab) domain. ZBTB5 was identified though sequence analysis as a POZ domain Kruppel-like zinc finger (POK) protein. Further experiments indicated that it binds DNA and can directly repress transcription of the cell cycle arrest gene p21. ZBTB5 can also interact with co-repressor histone deacetylase complexes such as BCoR, NCoR, and SMRT via its POZ domain, resulting in deacetylation of histones Ac-H3 and Ac-H4 at the proximal promoter. ZBTB5 stimulates both cell proliferation and cell cycle progression, suggesting that it may act as a potential proto-oncogene.
Type: Primary
Antigen: ZBTB5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat