Order Entry
United States
ContactUsLinkComponent
Lactate Dehydrogenase Activity Colorimetric Assay Kit, BioVision
Lactate Dehydrogenase Activity Colorimetric Assay Kit, BioVision
Catalog # 10186-988
Supplier:  Avantor
CAS Number:  
undefined
Lactate Dehydrogenase Activity Colorimetric Assay Kit, BioVision
Catalog # 10186-988
Supplier:  Avantor
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Size:
    500 assays
  • Environmentally Preferable:
  • Species Reactivity:
    Mammalian samples
  • Shelf Life:
    12 Months
  • Sample Size:
    Cell and tissue culture supernatants, urine, plasma and serum, as well as many other biological fluids etc.
  • Storage Temperature:
    -20°C
  • Cat. No.:
    10186-988
  • Supplier no.:
    K726-500

Specifications

About this item

Detection method- Absorbance (450 nm)
Species reactivity- Mammalian
Application- The kit can detect 1 -100 mU/ml of LDH directly in samples.

Lactate dehydrogenase (LDH) is an oxidoreductase (EC 1.1.1.27) present in a wide variety of organisms. It catalyses the interconversion of pyruvate and lactate with concomitant interconversion of NADH and NADᶧ. When disease or injury or toxic material damages tissues, cells release LDH into the bloodstream. Since LDH is a fairly stable enzyme, LDH has been widely used to evaluate the presence of damage and toxicity of tissue and cells. Quantification of LDH has broad range of applications. In this colorimetric LDH quantification assay, LDH reduces NAD to NADH, which then interacts with a specific probe to produce a color (λmax = 450 nm). The kit quantifies LDH activity in variety of biological samples such as in serum or plasma, cells, culture medium and fermentation, etc. The assay is quick, convenient, and sensitive. The kit can detect 1 -100 mU/ml of LDH directly in samples.