Order Entry
United States
ContactUsLinkComponent
Lipase Activity Colorimetric Assay Kit II, BioVision
Lipase Activity Colorimetric Assay Kit II, BioVision
Catalog # 10186-982
Supplier:  Avantor
CAS Number:  
undefined
Lipase Activity Colorimetric Assay Kit II, BioVision
Catalog # 10186-982
Supplier:  Avantor
CAS Number:  

Specifications

  • Size:
    100 assays
  • Species Reactivity:
    Mammalian samples
  • Shelf Life:
    12 Months
  • Sample Size:
    Cell and tissue culture supernatants, urine, plasma and serum, as well as many other biological fluids, fermentation media, food samples, etc.
  • Storage Temperature:
    -20°C
  • Cat. No.:
    10186-982
  • Supplier no.:
    K723-100

Specifications

About this item

Sensitive Assay, HTS

Detection method- Absorbance (412 nm)
Species reactivity- Mammalian

Lipase is a subclass of the esterases that catalyze the hydrolysis of ester bonds in water-insoluble, lipid substrates. Lipases perform essential roles in the digestion, transport and processing of dietary lipids (e.g. triglycerides, fats, oils) in most, if not all, living organisms. In humans, pancreatic lipases are the key enzyme responsible for breaking down fats in the digestive system by converting triglycerides to monoglycerides and free fatty acids. During the damage of the pancreas, lipase levels can rise 5 to 10-fold within 24 to 48 hours. The kit provides a simple, sensitive, and reliable assay for rapid analysis of Lipase in samples. In the assay, Lipases hydrolyze a specific substrate to generate a product which reacts with the DTNB probe to generate color (λ=412 nm). The kit is also suitable for high throughput analyses.