Specifications
- Size:100 assays
- Species Reactivity:Mammalian samples
- Shelf Life:12 Months
- Sample Size:Purified/Isolated enzyme, Testing inhibitor panels
- Storage Temperature:-20°C
- Cat. No.:10186-452
- Supplier no.:K709-100
Specifications
About this item
Sensitive Assay, HTS
Detection method- Absorbance (570 nm) or Fluorescence (Ex/Em 535/587 nm)
Species reactivity- Mammalian
Applications- The kit detects 0.1 mU/ml pyruvate kinase.
Pyruvate kinase (PK, EC 2.7.1.40) is an enzyme involved in glycolysis. It catalyzes the transfer of a phosphate group from phosphoenolpyruvate (PEP) to ADP, yielding one molecule of pyruvate and one molecule of ATP. Lack of pyruvate kinase will slow down the process of glycolysis which causes the disease known as pyruvate kinase deficiency. BioVision provides a simple, direct and automation-ready procedure for measuring pyruvate kinase activity in various biological samples such as blood, tissues, and culture cells, etc. In the assay, PEP and ADP were catalyzed by PK to generate pyruvate and ATP. The generated pyruvate is oxidized by pyruvate oxidase to produce color (at λ = 570 nm) and fluorescence (at Ex/Em = 535/587 nm). Since the increase in color or fluorescence intensity is proportional to the increase in pyruvate amount, the PK activity can be accurately measured. The kit detects 0.1 mU/ml pyruvate kinase.