Order Entry
United States
ContactUsLinkComponent
Glucose Uptake Colorimetric Assay Kit, BioVision
Glucose Uptake Colorimetric Assay Kit, BioVision
Catalog # 10191-570
Supplier:  Avantor
CAS Number:  
undefined
Glucose Uptake Colorimetric Assay Kit, BioVision
Catalog # 10191-570
Supplier:  Avantor
CAS Number:  

Specifications

  • Size:
    100 assays
  • Species Reactivity:
    Mammalian samples
  • Shelf Life:
    12 Months
  • Sample Size:
    Living cells (suspension and adherent)
  • Storage Temperature:
    -20°C
  • Cat. No.:
    10191-570
  • Supplier no.:
    K676-100

Specifications

About this item

Non-radioactive, Highly Sensitive Assay

Detection method- Absorbance (412 nm)
Species reactivity- Mammalian
Application- This easy to use non-radioactive kit is highly sensitive and can detect glucose uptake as low as 10 pmol/well.

Glucose uptake is an important biological process for studying cell signaling and glucose metabolism. Among many different methods available for measuring glucose uptake, 2-deoxyglucose (2-DG) has been widely used because of its structural similarity to glucose. As with glucose, 2-DG can be taken up by glucose transporters and metabolized to 2-DG-6-phosphate (2-DG6P). 2-DG6P, however, cannot be further metabolized, and thus accumulates in the cells. The accumulated 2-DG6P is directly proportional to 2-DG (or glucose) uptake by cells. In BioVision’s glucose uptake colorimetric assay kit, the 2-DG6P is oxidized to generate NADPH, which can be determined by an enzymatic recycling amplification reaction. This easy to use non-radioactive kit is highly sensitive and can detect glucose uptake as low as 10 pmol/well.