Order Entry
United States
ContactUsLinkComponent
Hemin Colorimetric Assay Kit, BioVision
Hemin Colorimetric Assay Kit, BioVision
Catalog # 10191-568
Supplier:  Avantor
CAS Number:  
undefined
Hemin Colorimetric Assay Kit, BioVision
Catalog # 10191-568
Supplier:  Avantor
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Size:
    100 assays
  • Species Reactivity:
    Mammalian samples
  • Shelf Life:
    12 Months
  • Sample Size:
    Cell and tissue culture supernatants, urine, plasma and serum, as well as many other biological fluids, growth media and food products
  • Storage Temperature:
    4°C
  • Cat. No.:
    10191-568
  • Supplier no.:
    K672-100

Specifications

About this item

Highly Sensitive Assay

Detection method- Absorbance (570 nm)
Species reactivity- Mammalian
Application- Trace amounts of Hemin can be quantitated in the 5-160 pg (10-250 fmol) range.

Free hemin results from the breakdown of hemin-containing proteins such as hemoglobin and myoglobin. It can be detected in various body fluids such as saliva, urine and csf under various pathological states. Free hemin exists in cells at a very minute concentrations (< 1µM ≈ 650 ng/ml) exerting regulatory functions such as repression of nonspecific δ-aminolevulinate synthase expression and induction of microsomal hemin oxygenase-1. Hemin can stimulate growth of oral bacteria involved in gingivitis and is an indicator of possible pathological conditions when found in the urine or feces. BioVision’s Hemin Assay Kit utilizes peroxidase activity in the presence of hemin to provide a simple, exquisitely sensitive assay which causes the conversion of a colorless probe to a strongly colored (λ = 570) compound. Trace amounts of Hemin can be quantitated in the 5-160 pg (10-250 fmol) range.