Free Glycerol Colorimetric/Fluorometric Assay Kit, BioVision
10186-896
: Avantor
:
- Size:100 assays
- Species Reactivity:Mammalian samples
- Shelf Life:12 Months
- Sample Size:Animal tissues & cells, Cell and tissue culture supernatants, plasma and serum, as well as many other biological fluids
- Storage Temperature:-20°C
- Cat. No.:10186-896
- Supplier no.:K630-100
Detection method- Absorbance (570 nm) or Fluorescence (Ex/Em 535/587 nm)
Applications-The kit can detect 50 pmol-10 nmol (or 1~10000 µM range) of glycerol in various samples.
Glycerol is widely used in foods, beverages, solvents, pharmaceutical and cosmetic products, etc. There is broad interest in quantification of glycerol for research and development. BioVision’s Glycerol Assay Kit provides a sensitive, easy assay to measure free glycerol concentration in various samples. In the assay, glycerol is enzymatically oxidized to generate a product which reacts with the probe to generate color (λ= 570 nm) and fluorescence (Ex/Em = 535/587 nm). The kit can detect 50 pmol-10 nmol (or 1~10000 µM range) of glycerol in various samples.