Order Entry
United States
Orders LinkContactUsLinkComponent
Triglyceride Quantification Colorimetric/Fluorometric Kit, BioVision

Triglyceride Quantification Colorimetric/Fluorometric Kit, BioVision

100 assays
Catalog # 10186-880
Triglyceride Quantification Colorimetric/Fluorometric Kit, BioVision
Catalog # 10186-880
Supplier:  
CAS Number:  
Triglyceride Quantification Colorimetric/Fluorometric Kit, BioVision
Triglyceride Quantification Colorimetric/Fluorometric Kit, BioVision

100 assays

CAS Number:  
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
SERUM TRIGLYCERIDE KIT FLUOR Catalog #102965-080

Product 102965-080 is similar to 10186-880 but manufacturer and specifications may vary.

SERUM TRIGLYCERIDE KIT COLOR Catalog #102965-078

Product 102965-078 is similar to 10186-880 but manufacturer and specifications may vary.

  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Detection method- Absorbance (570 nm) or Fluorescence (Ex/Em 535/587 nm)
Species reactivity- Mammalian
Applications- The kit can detect 2 pmol-10nmol (or 2-10000µM range) of triglyceride in various samples. The kit also detects monoglycerides and diglycerides.

Triglycerides (TG) are the main constituent of vegetable oil, animal fat, LDL and VLDL, and play an important role as transporters of fatty acids as well as serving as an energy source. TG are broken down into fatty acids and glycerol, after which both can serve as substrates for energy producing and metabolic pathways. High blood levels of TG are implicated in atherosclerosis, heart disease and stroke as well as in pancreatitis.The Triglyceride Quantification Kit provides a sensitive, easy assay to measure TG concentration in a variety of samples. In the assay, TG are converted to free fatty acids and glycerol. Theglycerol is then oxidized to generate a product which reacts with theprobe to generate color (spectrophotometry at λ = 570 nm) and fluorescence (Ex/Em = 535/587 nm). The kit can detect 2 pmol-10nmol (or 2-10000µM range) of triglyceride in various samples. The kit also detects monoglycerides and diglycerides.

Specifications

Specifications

  • Size:
    100 assays
  • Cat. No.:
    10186-880
  • Sample Size:
    Cell and tissue culture supernatants, urine, plasma, serum, as well as many other biological fluids
  • Species Reactivity:
    Mammalian samples
  • Shelf Life:
    12 Months
  • Storage Temperature:
    -20°C

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.