Cannon-Manning Semi-Micro Extra Low Charge Viscometer, Cannon Instrument
Supplier: Cannon
Certificates
About this item
The Cannon-Manning Semi-Micro Viscometer measures Available calibrated (CMSMC) and uncalibrated (CMSMU)
Minimum sample volume approximately 0.5 mL. Minimum bath depth 8".
Frequently Bought Together