Order Entry
United States
ContactUsLinkComponent
Pyruvate Colorimetric/Fluorometric Assay Kit, BioVision
Pyruvate Colorimetric/Fluorometric Assay Kit, BioVision
Catalog # 10186-856
Supplier:  Avantor
CAS Number:  
undefined
Pyruvate Colorimetric/Fluorometric Assay Kit, BioVision
Catalog # 10186-856
Supplier:  Avantor
CAS Number:  

Specifications

  • Size:
    100 assays
  • Species Reactivity:
    Mammalian samples
  • Shelf Life:
    12 Months
  • Sample Size:
    Animal tissues, Cell culture: adherent or suspension cells and Serum, saliva
  • Storage Temperature:
    -20°C
  • Cat. No.:
    10186-856
  • Supplier no.:
    K609-100

Specifications

About this item

Simple Assay, HTS

Detection method- Absorbance (570 nm) or Fluorescence (Ex/Em 535/587 nm)
Species reactivity- Mammalian
Application- The kit detects 1 µM to 10 mM pyruvate.

Pyruvate is a central molecule in metabolism through which sugars enter the citric acid cycle. Pyruvate can be converted to carbohydrates during gluconeogenesis or to fatty acids via acetyl CoA. High levels of pyruvate are associated with liver disease and genetic disorders. Pyruvate has also been used to stimulate metabolism leading to loss of body weight. BioVision provides a simple, direct and automation-ready procedure for measuring pyruvate concentration in various biological samples such as blood, cells, culture and fermentation media, etc. In the assay, pyruvate is oxidized by pyruvate oxidase via enzyme reactions to generate color (λ= 570 nm) and fluorescence (at Ex/Em = 535/587 nm). Since the color or fluorescence intensity is proportional to pyruvate content, the pyruvate concentration can be accurately measured. The kit detects 1 µM to 10 mM pyruvate.