Order Entry
United States
ContactUsLinkComponent
Total Cholesterol and Cholesteryl Ester Colorimetric/Fluorometric Assay Kit, BioVision
Total Cholesterol and Cholesteryl Ester Colorimetric/Fluorometric Assay Kit, BioVision
Catalog # 10191-550
Supplier:  Avantor
CAS Number:  
undefined
Total Cholesterol and Cholesteryl Ester Colorimetric/Fluorometric Assay Kit, BioVision
Catalog # 10191-550
Supplier:  Avantor
CAS Number:  

Specifications

  • Size:
    100 assays
  • Species Reactivity:
    Mammalian samples
  • Shelf Life:
    12 Months
  • Sample Size:
    Cell and tissue culture supernatants, urine, plasma, serum, as well as many other biological fluids
  • Storage Temperature:
    -20°C
  • Cat. No.:
    10191-550
  • Supplier no.:
    K603-100

Specifications

About this item

Detection method- Absorbance (570 nm) or Fluorescence (Ex/Em 535/587 nm)
Species reactivity- Mammalian
Application- The assay detects total cholesterol (cholesterol and cholesteryl esters) in the presence of cholesterol esterase or free cholesterol in the absence of cholesterol esterase in the reaction.

The Cholesterol/Cholesteryl Ester Quantitation Kit provides a simple method for sensitive quantification of free cholesterol, cholesteryl esters, or both by colorimetric or fluorometric methods. Majority of the cholesterol in blood is in the form of cholesteryl esters which can be hydrolyzed to cholesterol by cholesterol esterase. Cholesterol is then oxidized by cholesterol oxidase to yield H₂O₂which reacts with a sensitive cholesterol probe to produce color (λmax = 570 nm) and fluorescence (Ex/Em = 535/587 nm). The assay detects total cholesterol (cholesterol and cholesteryl esters) in the presence of cholesterol esterase or free cholesterol in the absence of cholesterol esterase in the reaction. Cholesteryl ester can be determined by subtracting the value of free cholesterol from the total (cholesterol plus cholesteryl esters).