Order Entry
United States
ContactUsLinkComponent
Sialic Acid (NANA) Colorimetric/Fluorometric Assay Kit, BioVision
Sialic Acid (NANA) Colorimetric/Fluorometric Assay Kit, BioVision
  10186-834
 :  Avantor
 :  
undefined
Sialic Acid (NANA) Colorimetric/Fluorometric Assay Kit, BioVision
  10186-834
 :  Avantor
 :  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

 

  • Size:
    100 assays
  • Environmentally Preferable:
  • Species Reactivity:
    Mammalian samples
  • Shelf Life:
    12 Months
  • Sample Size:
    Cell and tissue culture supernatants, urine, plasma, serum, as well as many other biological fluids
  • Storage Temperature:
    -20°C
  • Cat. No.:
    10186-834
  • Supplier no.:
    K566-100

 

 

Detection method- Absorbance (570 nm) or Fluorescence (Ex/Em 535/587 nm)
Species reactivity- Mammalian
Applications- The kit measures sialic acid in the linear range of 0.1 to 10 nmol with a detection sensitivity ~1 µM concentration.

Sialic acid is a generic term for the N- or O-substituted derivatives of neuraminic acid, a monosaccharide with a nine-carbon backbone. It is also the name for the most common member of this group, N-acetylneuraminic acid. Sialic acids are found widely distributed in animal tissues and to a lesser extent in other species ranging from plants and fungi to yeasts and bacteria, mostly in glycoproteins and gangliosides. It has been shown recently that sialic acid level may be associated with developmental and pathological stages. BioVision's Sialic Acid Assay Kit provides a simple and convenient means of measuring free Sialic Acid in a variety of biological samples. The kit utilizes an enzyme coupled reaction in which free sialic acid is oxidized resulting in development of the Oxi-Red probe to give fluorescence (Ex/Em=535/587 nm) and absorbance (O.D.=570 nm). The kit measures sialic acid in the linear range of 0.1 to 10 nmol with a detection sensitivity ~1 µM concentration.