Hydroxyproline Colorimetric Assay Kit, BioVision
Catalog # 10186-830
Supplier: Avantor
CAS Number:
Specifications
- Species Reactivity:Mammalian samples
- Shelf Life:12 Months
- Sample Size:Animal tissues, protein/peptide hydrolysates, serum and urine
- Storage Temperature:+4°C