Cannon-Ubbelohde Viscometer, Cannon Instrument
Supplier: Cannon
Certificates
About this item
The Cannon-Ubbelohde Viscometer, with a more rugged design than standard Ubbelohde viscometers, measures kinematic viscosity of transparent Newtonian liquids according to ASTM D445 and ISO 3104
Available calibrated (CUC) and uncalibrated (CUU). Minimum sample volume 11 mL. Minimum bath depth 10".
Frequently Bought Together