Order Entry
United States
ContactUsLinkComponent
Phosphate Colorimetric Assay Kit, BioVision
Phosphate Colorimetric Assay Kit, BioVision
Catalog # 10186-692
Supplier:  Avantor
CAS Number:  
undefined
Phosphate Colorimetric Assay Kit, BioVision
Catalog # 10186-692
Supplier:  Avantor
CAS Number:  

Specifications

  • Size:
    500 assays
  • Species Reactivity:
    Mammalian samples
  • Shelf Life:
    12 Months
  • Sample Size:
    Cell and tissue lysates, culture media, urine, plasma and serum, as well as many other biological fluids, algal blooms and water from run-off areas of high fertilizer use
  • Storage Temperature:
    RT
  • Cat. No.:
    10186-692
  • Supplier no.:
    K410-500

Specifications

About this item

Detection method- Absorbance (650 nm)
Species reactivity- Mammalian
Applications- Phosphate concentrations between 1 µM and 1 mM, with a lower limit of detection of approximately 0.1 nmol, can be directly determined.

Phosphate is one of the most important of the inorganic ions in biological systems. It functions in a variety of roles. One of the most important roles is as a molecular switch, turning enzyme activity on and off through the mediation of the various protein kinases and phosphatases in biological systems. Phosphate is also of great importance in mineralization processes and is a primary stimulus of algal blooms frequently found in bodies of fresh water, due to run-off from areas of high fertilizer use. The newly designed Phosphate Colorimetric Assay Kit provides an easy, quick and sensitive means of assessing phosphate over a wide range of concentrations. The assay utilizes a proprietary formulation of malachite green and ammonium molybdate which forms a chromogenic complex with phosphate ion giving an intense absorption band around 650 nm. Phosphate concentrations between 1 µM and 1 mM, with a lower limit of detection of approximately 0.1 nmol, can be directly determined. The Phosphate Colorimetric Assay Kit provides 500 assays using microtiter plates or 100 assays using 1 ml cuvettes.