Order Entry
United States
ContactUsLinkComponent
Senescence Detection Kit, BioVision
Senescence Detection Kit, BioVision
Catalog # 10186-622
Supplier:  Avantor
CAS Number:  
undefined
Senescence Detection Kit, BioVision
Catalog # 10186-622
Supplier:  Avantor
CAS Number:  

Specifications

  • Size:
    250 Stainings
  • Species Reactivity:
    Mammalian samples
  • Shelf Life:
    12 Months
  • Sample Size:
    Cell culture cells, tissues
  • Storage Temperature:
    -20°C
  • Cat. No.:
    10186-622
  • Supplier no.:
    K320-250

Specifications

About this item

Histochemical kit

Species reactivity- Mammalian
Kit size- 250 stainings
Applications- The Senescence Detection Kit is designed to histochemically detect specific senescence marker in distinct pH in cultured cells and tissue sections.

Senescence is thought to be a tumor suppressive mechanism and an underlying cause of aging. Senescence represents an arrested state in which the cells remain viable, but not stimulated to divide by serum or passage in culture. Senescent cells display increase of cell size, senescence-associated expression of β-galactosidase (SA-β-Gal) activity, and altered patterns of gene expression. The Senescence detection kit is designed to histochemically detect SA-β-Gal activity in cultured cells and tissue sections, a known characteristic of senescent cells. The SA-β-Gal is present only in senescent cells and is not found in presenescent, quiescent or immortal cells.