To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Size:500 assays
- Species Reactivity:Mammalian samples
- Shelf Life:12 Months
- Sample Size:Cell cultures (adherent and suspension)
- Storage Temperature:-20°C
- Cat. No.:10186-616
- Supplier no.:K313-500
Specifications
About this item
Highly sensitive assay
Detection method- Absorbance (450 nm)
Species reactivity- Mammalian
Applications- This assay provides a convenient and Wst-based non-radioactive assay for measurement of activity of lactate dehydrogenase (LDH) which is a stable enzyme normally found in the cytosol of all cells but rapidly releases into the supernatant upon damage of plasma membrane.
Cell death or cytotoxicity is classically evaluated by the quantification of plasma membrane damage. Lactate dehydragenase (LDH) is a stable enzyme, present in all cell types, and rapidly released into the cell culture medium upon damage of the plasma membrane. LDH, therefore, is the most widely used marker in cytotoxicity study. BioVision’s LDH Cytotoxicity Assay Kit II utilizes the advanced WST reagent for a fast and more sensitive detection of LDH released from damaged cells. The assay utilizing an enzymatic coupling reaction: LDH oxidizes lactate to generate NADH, which then reacts with WST to generate yellow color. The intensity of the generated color correlates directly with the cell number lysed. Since WST is brighter, less amount of culture medium is required for the assay, and thus the background from serum and culture medium is significantly reduced. Using the assay, cells can be cultured in regular 10% serum containing medium, no reducing serum or special medium is required for the assay. In addition, since the WST is more stable, the reaction can be read multiple times, and can also be stopped at any time point during the reaction. LDH activity can be easily quantified by spectrophotometer or plate reader at OD450 nm. The kit provides all necessary reagents including LDH positive control. Assay takes less than 1 hour.