Order Entry
United States
ContactUsLinkComponent
Quick Cell Proliferation Colorimetric Assay Kit, BioVision Plus
Quick Cell Proliferation Colorimetric Assay Kit, BioVision Plus
Supplier:  Avantor
Certificates
undefined
Quick Cell Proliferation Colorimetric Assay Kit, BioVision Plus
Supplier:  Avantor

Specifications

  • Species Reactivity:
    Mammalian samples
  • Shelf Life:
    12 Months
  • Sample Size:
    Cell cultures (adherent and suspension)
  • Storage Temperature:
    -20°C

Specifications

Related Information

Frequently Bought Together