Order Entry
United States
ContactUsLinkComponent
Yeast Nuclei Isolation Kit, BioVision
Yeast Nuclei Isolation Kit, BioVision
Catalog # 10186-444
Supplier:  Avantor
CAS Number:  
undefined
Yeast Nuclei Isolation Kit, BioVision
Catalog # 10186-444
Supplier:  Avantor
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Size:
    50 assays
  • Species Reactivity:
    Mammalian samples
  • Shelf Life:
    12 Months
  • Sample Size:
    Yeast cell culture
  • Storage Temperature:
    -20°C
  • Cat. No.:
    10186-444
  • Supplier no.:
    K289-50

Specifications

About this item

Only kit on the market to isolate yeast nuclei

Application:
DNA-Protein interaction, RNA-Protein interaction and Protein-Protein interaction studies
DNase I footprinting analysis, Enzymatic Assays and Pull-down assay
Western blot and ELISA

To understand the nature of the replication and expression of the yeast genome, it is essential to have a method for the preparation of nuclei at different stages of growth. BioVision’s Yeast Nuclei Isolation kit enables fast and easy purification of nuclei from yeast cells, utilizing yeast cell wall lysis and homogenization.