Order Entry
United States
ContactUsLinkComponent
Mitochondrial DNA Isolation Kit, BioVision
Mitochondrial DNA Isolation Kit, BioVision
Catalog # 10186-596
Supplier:  Avantor
CAS Number:  
undefined
Mitochondrial DNA Isolation Kit, BioVision
Catalog # 10186-596
Supplier:  Avantor
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Size:
    50 assays
  • Environmentally Preferable:
  • Species Reactivity:
    Mammalian samples
  • Shelf Life:
    12 Months
  • Sample Size:
    Cell cultures (adherent and suspension), fresh and frozen tissues
  • Storage Temperature:
    -20°C
  • Cat. No.:
    10186-596
  • Supplier no.:
    K280-50

Specifications

About this item

Isolates mt

Species reactivity- Mammalian
Applications- Enzyme manipulations, Southern blotting, cloning, PCR analysis, and amplifications.

Mitochondria are semiautonomous organelles which functions in aging process, apoptosis, anti-HIV drugs, and cancers. Mitochondrial DNA (mtDNA) has a very high mutation rate and the mutations on mtDNA appear to be related to certain diseases such as diabetes, Alzheimer’s disease, and muscle disorders. Isolation and quantification of mtDNA are often required to study the relationships between the diseases and mtDNA. The Mitochondrial DNA Extraction Kit provides convenient tools for isolating mtDNA from a variety of cells and tissues in high yield and purity, without contaminations from genomic DNA. The purified mtDNA can be used for a variety of studies such as enzyme manipulations, Southern blotting, cloning, PCR analysis, and amplifications.

DNA, no genomic DNA contamination