Isolates nuclear/cytosol fractions w/o untracentrifugation
Species reactivity- Mammalian
Kit size- Conveient sizes (25, 100)
Applications- This Nuclear/Cytosol Extraction Kit provides a complete system that enables the separation of nuclear extract from the cytoplasmic fraction of mammalian cells and tissues The new kit provides a system that maintains the nuclear and cytoplasmic compartments separate and intact. Transcriptional activity/reporter assays/enzyme activity assays/Western blotting and more downstream assays.
This Nuclear/Cytosol Extraction Kit provides a complete system that enables the separation of nuclear extract from the cytoplasmic fraction of mammalian cells. The optimized reagents and procedures provided with the kit allow separation of nuclear and cytoplasmic fractions quickly with little or no cross-contaminations. The extracted nuclear and cytoplasmic protein fractions are functional and compatible with downstream assays such as transcriptional activity, RNA splicing, gel shift assay, reporter assays, enzyme activity assays, and Western blotting.
- Species Reactivity:Mammalian samples
- Shelf Life:12 Months
- Sample Size:Cell cultures (adherent and suspension), fresh and frozen tissues
- Storage Temperature:-20°C