Order Entry
United States
ContactUsLinkComponent
Cytochrome c Releasing Apoptosis Assay Kit, BioVision
Cytochrome c Releasing Apoptosis Assay Kit, BioVision
Catalog # 10186-568
Supplier:  Avantor
CAS Number:  
undefined
Cytochrome c Releasing Apoptosis Assay Kit, BioVision
Catalog # 10186-568
Supplier:  Avantor
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Size:
    100 assays
  • Species Reactivity:
    Mammalian samples
  • Shelf Life:
    12 Months
  • Sample Size:
    Cells and tissues
  • Storage Temperature:
    -20°C
  • Cat. No.:
    10186-568
  • Supplier no.:
    K257-100

Specifications

About this item

Detection method- Western blotting
Species reactivity- Mammalian
Applications- Effective means for detecting cytochrome c translocation from mitochondria into cytosol during apoptosis. The kit provides an easy-to-use procedure and reagents for separating a mitochondria-enriched fraction from cytosol.

Cytochrome c plays an important role in apoptosis. The protein is located in the space between the inner and outer mitochondrial membranes. An apoptotic stimulus triggers the release of cytochrome c from the mitochondria into cytosol where it binds to Apaf-1. The cytochrome c/Apaf-1 complex activates caspase-9, which then activates caspase-3 and other downstream caspases. BioVision’s Cytochrome c Releasing Apoptosis Assay Kit provides an effective means for detecting cytochrome c translocation from mitochondria into cytosol during apoptosis. The kit provides unique formulations of reagents to isolate a highly enriched mitochondria fraction from cytosol. The procedure is so simple and easy to perform, no ultracentrifugation is required and no toxic chemicals are involved. Cytochrome c releasing from mitochondria into cytosol is then determined by Western blotting using the cytochrome c antibody provided in the kit.