CaspGLOW™ Fluorescein Active Caspase-9 Staining Kit, BioVision
Supplier: Avantor
Certificates
About this item
Detects activated Caspase-9 by FACS, FL or plate reader
Detection method- Fluorescence microscopy, Flow cytometry and fluorescence plate reader (Ex. = 485 nm and Em. = 535 nm)
Species reactivity- Mammalian
Kit size- Convenient sizes (100 assays)
Applications- Sensitive detection of activated caspase-9 in living cells.
Activation of caspases plays a central role in apoptosis. The CaspGLOW™ Fluorescein Active Caspase-9 Staining Kit provides a convenient means for sensitive detection of activated caspase-9 in living cells. The assay utilizes the caspase-9 inhibitor LEHD-FMK conjugated to FITC (FITC-LEHD-FMK) as a fluorescent marker. FITC-LEHD-FMK is cell permeable, nontoxic, and irreversibly binds to activated caspase-9 in apoptotic cells.
Specifications
- Species Reactivity:Mammalian samples
- Shelf Life:6-12 months
- Sample Size:Live cells
- Storage Temperature:-20°C
Specifications
Frequently Bought Together