About this item
Convenient, Highly-Sensitive Assay
Detection method- Absorbance (400 or 405 nm)
Species reactivity- Mammalian
Kit size- Convenient sizes (25, 100, 200, 400 assays)
Applications- Detect early/middle stages of apoptosis; differentiate apoptosis from necrosis.
Activation of ICE-family proteases/caspases initiates apoptosis in mammalian cells. The Caspase-3/CPP32 Colorimetric Assay Kit provides a simple and convenient means for assaying the activity of caspases that recognize the sequence DEVD. The assay is based on spectrophotometric detection of the chromophore p-nitroaniline (pNA) after cleavage from the labeled substrate DEVD-pNA. The pNA light emission can be quantified using a spectrophotometer or a microtiter plate reader at 400- or 405 nm. Comparison of the absorbance of pNA from an apoptotic sample with an uninduced control allows determination of the fold increase in CPP32 activity.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Species Reactivity:Mammalian samples
- Shelf Life:12 Months
- Sample Size:Cell and tissue lysates
- Storage Temperature:-20°C