- Description:Hematek® 3000 Semi Automated Slide Stainer with Dryer
- Cat. No.:10124-406
- Electrical:110 - 230 V
- D×W×H:46×54×22 cm (18.1×21.3×8.7")
- Weight:20 kg (44 lbs.)
- Supplier no.:10805311
Hematek® 3000 Hematology System from Siemens Healthcare Diagnostics provides semiautomated slide staining for laboratories that need a robust system capable of staining up to 60 slides per hour.
- Continuous-feed loading platform
- Minimal system maintenance
- Easy load-and-go operation
- Minimal hands-on time required
Built on a heritage of slide staining spanning more than half a century, the system helps maximize productivity and deliver quality results through user-defined features and precision controls
The Hematek® 3000 System is designed to complement the Siemens portfolio of hematology analyzers and digital microscopy solutions. The Hematek 3000 System is built on the proven technology of its predecessors, while new features further support easy and environmentally friendly features. All-in-one stain pack includes enough stain for up to 900 slides and offers easy load-and-go operation for optimized daily workflow with minimal hands-on time. Pumps deliver a fresh, precise volume of reagent for every slide, while the platen provides uniform stain from field to field and slide to slide with accurate mixing and uniform stain and buffer delivery. Simple pump release mechanism streamlines maintenance. Slides are immediately available for viewing upon completion of staining, and the rinse reagent contains the optimal alcohol concentration for proper slide drying. System warnings alert users of low stain-level and full waste bin to help ensure uninterrupted, quality results.
Continuous-feed loading platform eliminates wasted time associated with batch staining methods. Individual, consistent slide staining optimizes staining quality and enables user-adaptable intensity for standardized slide staining. Minimal system maintenance helps maximize uptime in your lab. Compact, RoHS-compliant system guarantees incorporation of environmentally friendly, sustainable substances and supports “green” initiatives.