ContactUsLinkComponent
KidWind Motor Mount Assembly
  470165-148
undefined
KidWind Motor Mount Assembly
  470165-148
 :  KW-MTRMNT
  • Description:
    KidWind Motor Mount Assembly
  • Cat. No.:
    470165-148
  • Supplier no.:
    KW-MTRMNT
The KidWind Motor Mount Pack holds the motor (generator) in place when affixed to the wind turbine nacelle. The Motor Mount Pack can be used to replace lost parts from a nacelle. It can also be used to mount a nacelle to the tower for a vertical axis wind turbine configuration.