ContactUsLinkComponent
Gear Set with Spool
Gear Set with Spool
Catalog # 470165-140
undefined
Gear Set with Spool
Catalog # 470165-140
Supplier Number:  KW-GEAR
Specifications
  • Description:
    Gear Set with Spool
  • Cat. No.:
    470165-140
  • Supplier no.:
    KW-GEAR
Specifications
About this item
Experiment with gear ratios on your wind turbine or other small projects. Adding gears to your turbine can greatly increase the generator RPM, giving you higher power output. The small 8-tooth gear fits on 2 mm driveshafts that are found on many DC generators. The gears have a keying feature that allows them to be changed quickly and easily using the included hex locks. The hex lock secures to our 9/64 inch hex-driveshaft. Hex-driveshaft not included.