Order Entry
United States
ContactUsLinkComponent
Human Recombinant R Spondin 1 (from HEK293 Cells)
Human Recombinant R Spondin 1 (from HEK293 Cells)
Catalog # 10797-888
Supplier:  Prosci
CAS Number:  
undefined
Human Recombinant R Spondin 1 (from HEK293 Cells)
Catalog # 10797-888
Supplier:  Prosci
Supplier Number:  96-660
CAS Number:  

Specifications

  • Protein/Peptide Type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Size:
    0.05 mg
  • Tag sequence:
    His Tag
  • Protein Synonyms:
    CRISTIN3|R-spondin-1|RP11-566C13.1|FLJ40906|RSPO1
  • Protein/Peptide Name:
    R Spondin 1
  • Purity:
    >95% as determined by SDS-PAGE.
  • Molecular Weight:
    16.7 kDa
  • Sequence:
    21-146
  • Formulation:
    Lyophilized, PBS, pH7.4
  • Tested Applications:
    Western Blot
  • Cat. No.:
    10797-888
  • Supplier no.:
    96-660

Specifications

About this item

R-spondin-1 is also known as Roof plate-specific Spondin 1 (RSPO1) and cysteinerich and single thrombospondin domain containing protein 3 (Cristin 3), is a secreted protein which belongs to the R-Spondin family and encodes a secreted activator protein with two cystein-rich, furin-like domains and one thrombospondin type 1 domain. All Rspondins regulate Wnt/?-catenin signaling, but have distinct expression patterns. Like other R-Spondins, R-Spondin-1 contains two adjacent cysteinerich furinlike domains (aa 34-135) with one potential N-glycosylation site, followed by a thrombospondin (TSP1) motif (aa 147-207) and a region rich in basic residues (aa 211-263). Only the furinlike domains are needed for ?-catenin stabilization. A putative nuclear localization signal at the C-terminus may allow some expression in the nucleus. Potential isoforms of 200 and 236 aa have an alternate, shorter N-terminus or are missing aa 146-208, respectively. R-Spondin-1 is expressed in early development at the roof plate boundary and is thought to contribute to dorsal neural tube development. Human RSPO1 disruption results in a recessive syndrome characterized by XX sex reversal, palmoplantar hyperkeratosis and predisposition to squamous cell carcinoma of the skin. It has been shown that the complete female-to-male sex reversal is due to the absence of the testis-determining gene, SRY. R-Spondin-1 regulates Wnt/?-catenin by competing with the Wnt antagonist DKK1 for binding to the Wnt co receptors, Kremen and LRP6, reducing their DKK1 mediated internalization. Reports differ on whether R-spondin 1 binds LRP6 directly.