Order Entry
United States
ContactUsLinkComponent
Recombinant Gp120 (from HEK293 cells)
Recombinant Gp120 (from HEK293 cells)
Catalog # 10797-382
Supplier:  Prosci
CAS Number:  
undefined
Recombinant Gp120 (from HEK293 cells)
Catalog # 10797-382
Supplier:  Prosci
Supplier Number:  96-367
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Protein/Peptide Type:
    Recombinant
  • Source:
    HEK293 cells
  • Size:
    0.1 mg
  • Tag sequence:
    His Tag
  • Protein Synonyms:
    GP120|GP120-16055
  • Protein/Peptide Name:
    Gp120
  • Purity:
    >95% as determined by SDS-PAGE.
  • Molecular Weight:
    54 kDa
  • Formulation:
    Lyophilized, PBS, pH7.4
  • Tested Applications:
    Western Blot
  • Cat. No.:
    10797-382
  • Supplier no.:
    96-367

Specifications

About this item

Human Immunodeficiency Virus (HIV) can be divided into two major types, HIV type 1 (HIV-1) and HIV type 2 (HIV-2). HIV-1 is related to viruses found in chimpanzees and gorillas living in western Africa. HIV-2 is related to viruses found in sooty mangabeys. HIV-1 viruses may be further divided into groups. The HIV-1 group M viruses predominate and are responsible for the AIDS pandemic. Some of the HIV-1 group M subtypes are known to be more virulent or are resistant to different medications. HIV-2 viruses are thought to be less virulent and transmissible than HIV-1 M group viruses.
Envelope glycoprotein GP120 (or gp120) is the name of the glycoprotein which forms the spikes sticking out of a HIV virus particle. gp120 is essential for virus entry into cells as it plays a vital role in seeking out specific cell surface receptors for entry. Three gp120s, bound as heterodimers to a transmembrane glycoprotein, gp41, are thought to combine in a trimer to form the envelope spike, which is involved in virus-cell attachment. One half of the molecular weight of gp120 is due to the carbohydrate side chains (the "glyco-" in "glycoprotein"). These are sugar residues which form something almost like a sugar "dome" over the gp120 spikes. This dome prevents gp120 from being recognised by the human immune response. As the HIV virus and the human CD4 cell come together, the gp120 binding site "snaps open" at the last minute.The glycoprotein gp120 is anchored to the viral membrane, or envelope, via non-covalent bonds with the transmembrane glycoprotein, gp41. It is involved in entry into cells by binding to CD4 receptors, particularly helper T-cells. Binding to CD4 is mainly electrostatic although there are van der Waals interactions and hydrogen bonds.