Order Entry
United States
ContactUsLinkComponent
Human Recombinant TNFRSF7 (from HEK293 Cells)
Human Recombinant TNFRSF7 (from HEK293 Cells)
Catalog # 10797-970
Supplier:  Prosci
CAS Number:  
undefined
Human Recombinant TNFRSF7 (from HEK293 Cells)
Catalog # 10797-970
Supplier:  Prosci
Supplier Number:  96-133
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Protein/Peptide Type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Size:
    0.2 mg
  • Environmentally Preferable:
  • Tag sequence:
    C-Fc Tag
  • Biological Activity:
    Measured by its binding ability in a functional ELISA. Immobilized recombinant human CD27 protein at 2μg/mL (100μl/well) can bind biotinylated human CD70 with a linear range of 0. 25 - 16 ng/mL.
  • Protein Synonyms:
    Tp55|S152|T14|TNFRSF7|CD27|S152. LPFS2
  • Protein/Peptide Name:
    TNFRSF7
  • Purity:
    >95% as determined by SDS-PAGE.
  • Molecular Weight:
    45.9 kDa
  • Formulation:
    Lyophilized, 50 mM tris, 100 mM glycine, pH7.5
  • Tested Applications:
    Western Blot
  • Cat. No.:
    10797-970
  • Supplier no.:
    96-133

Specifications

About this item

CD Antigen CD27 is also known as Tumor necrosis factor receptor superfamily member 7 (TNFRSF7), which belongs to TNF-receptor superfamily. CD27 / TNFRSF7 is found in most T-lymphocytes. CD27 / TNFRSF7 is required for generation and long-term maintenance of T cell immunity. It binds to ligand CD70, and plays a key role in regulating B-cell activation and immunoglobulin synthesis. This receptor transduces signals that lead to the activation of NF-?B and MAPK8/JNK. CD27 has been shown to interact with SIVA1, TRAF2 and TRAF3.