Order Entry
United States
ContactUsLinkComponent
Human Recombinant IL2RB (from HEK293 Cells)
Human Recombinant IL2RB (from HEK293 Cells)
Catalog # 10797-138
Supplier:  Prosci
CAS Number:  
undefined
Human Recombinant IL2RB (from HEK293 Cells)
Catalog # 10797-138
Supplier:  Prosci
Supplier Number:  96-097
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Protein/Peptide Type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Size:
    0.02 mg
  • Environmentally Preferable:
  • Tag sequence:
    His Tag
  • Protein Synonyms:
    CD122|P70-75|IL2RB|RP5-1170K4.6
  • Protein/Peptide Name:
    IL2RB
  • Purity:
    >90% as determined by SDS-PAGE.
  • Molecular Weight:
    25.4 kDa
  • Formulation:
    Lyophilized, PBS, pH7.4
  • Tested Applications:
    Western Blot
  • Cat. No.:
    10797-138
  • Supplier no.:
    96-097

Specifications

About this item

Interleukin-2 receptor (IL-2R) is a heterotrimeric protein expressed on the surface of certain immune cells, such as lymphocytes, that binds and responds to a cytokine called IL-2. The IL-2R is made up of 3 subunits - ? (CD25), ? (CD122) and ? (CD132) - non-covalently associating. The ? and ? chains are involved in binding IL-2, while signal transduction following cytokine interaction is carried out by the ?-chain, along with the ? subunit.
CD122 is also known as IL2R beta, is a member of the type I cytokine receptor family. CD122 is the receptor for interleukin-2 and is involved in receptor mediated endocytosis and transduces the mitogenic signals of IL2.