CCDA Selective Supplement, MilliporeSigma
Catalog # EM1.00071.0010
Supplier: MilliporeSigma
CAS Number:
Specifications
- Pack type:Fiber case
- Description:CCDA Selective Supplement
- Size:10 x 1 vial
- Shelf Life:36 months from mfg date
- Storage:Store at +2°C to +8°C
- Cat. No.:EM1.00071.0010
- Supplier no.:1.00071.0010
Specifications
About this item
- 0.01% recovery on Escherichia coli ATCC 25922
- 70% minimum recovery on Campylobacter jejuni ATCC 29428
- 50% minimum recovery on Campylobacter jejuni ATCC 33291
- 70% minimum recovery on Campylobacter coli ATCC 33559
- 1% maximum recovery on Saccharomyces cerevisiae ATCC 9763