Sticky Mat Refills, Ergomat
About this item
Ergomat Sticky Mats provide a fast, convenient, reliable way to clean shoe soles or other items before entering a clean and sanitary area
- 30 layers per mat
- Easily adhered to a frame with double-faced adhesive tape
- Mats utilize a water-based acrylic adhesive, which has a pH of 7–8
- A clear release sheet protects the top sheet of the sticky mat until it is ready to use
- Corner tabs are numbered to show the number of remaining sheets
These significantly reduce traffic-borne contaminants and have proven themselves effective in a wide variety of industries such as microelectronics, aerospace, pharmaceuticals, and food processing. The disposable sticky mat consists of a stack of tear-off polyethylene films full of adhesive sheets that effectively capture dirt and dust.
Certifications: Meets DIN/ASTM requirements.
Ordering information: Frame must be ordered separately.
2 Options Available Below
Product Details & Documents
Ergomat Sticky Mats provide a fast, convenient, reliable way to clean shoe soles or other items before entering a clean and sanitary area
- 30 layers per mat
- Easily adhered to a frame with double-faced adhesive tape
- Mats utilize a water-based acrylic adhesive, which has a pH of 7–8
- A clear release sheet protects the top sheet of the sticky mat until it is ready to use
- Corner tabs are numbered to show the number of remaining sheets
These significantly reduce traffic-borne contaminants and have proven themselves effective in a wide variety of industries such as microelectronics, aerospace, pharmaceuticals, and food processing. The disposable sticky mat consists of a stack of tear-off polyethylene films full of adhesive sheets that effectively capture dirt and dust.
Certifications: Meets DIN/ASTM requirements.
Ordering information: Frame must be ordered separately.
Product Family Options
Product Information
- PackagingWidthLengthColorAvailabilityPrice
- 10067-686Sticky Mat RefillsSticky Mat Refills30 Sheets/Refill, 10 Refills/Box46 cm (18")114 cm (45")WhiteBox of 300$327.80Each$327.80Supplier Number: SM1504-WHITE
- 10067-688Sticky Mat RefillsSticky Mat Refills30 Sheets/Refill, 10 Refills/Box46 cm (18")114 cm (45")BlueLog in for availabilityEach$342.84Supplier Number: SM1504-BLU