Order Entry
United States
ContactUsLinkComponent
BAX® System Real-Time PCR Assay for Campylobacter jejuni / coli / lari, Hygiena™, Qualicon Diagnostics LLC
BAX® System Real-Time PCR Assay for Campylobacter jejuni / coli / lari, Hygiena™, Qualicon Diagnostics LLC
Catalog # 10754-888
Supplier:  Hygiena
CAS Number:  
undefined
BAX® System Real-Time PCR Assay for Campylobacter jejuni / coli / lari, Hygiena™, Qualicon Diagnostics LLC
Catalog # 10754-888
Supplier:  Hygiena
Supplier Number:  KIT2018
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Sample Type:
    Turkey and Poultry Carcass Rinses
  • Processing Time:
    70 minutes
  • Cat. No.:
    10754-888
  • Response time:
    24-48 hours
  • Target group:
    C. jejuni, C. coli, C. lari
  • Supplier no.:
    D12683449
  • No. of tests:
    96

Specifications

About this item

The Hygiena™ BAX® System PCR Assay for Campylobacter jejuni/coli/lari produces clear yes-or-no results after a fast processing time in the BAX® System Q7 instrument.

The Hygiena BAX System Real-Time PCR Assay for Campylobacter jejuni/coli/lari produces clear yes-or-no results for all three strains after 70 minutes of processing in the BAX System Q7 instrument. Highly contaminated samples can be tested directly, without enrichment, for even faster results. Certified as an AOAC-RI Performance Tested MethodSM for processed turkey and poultry carcass rinses.

The Hygiena BAX System is an automated detection system that uses the power of the polymerase chain reaction (PCR) to detect harmful bacteria and other microbes in food ingredients, finished products and environmental surfaces.

Ordering information: Includes PCR tubes with tablets, optical caps, protease, and lysis buffer. This assay can only be used with the BAX System Q7 instrument.