Order Entry
United States
ContactUsLinkComponent
BAX® System Real-Time PCR Assay for Vibrio cholerae / parahaemolyticus / vulnificus, Hygiena™, Qualicon Diagnostics LLC
BAX® System Real-Time PCR Assay for Vibrio cholerae / parahaemolyticus / vulnificus, Hygiena™, Qualicon Diagnostics LLC
Catalog # 10754-892
Supplier:  Hygiena
CAS Number:  
undefined
BAX® System Real-Time PCR Assay for Vibrio cholerae / parahaemolyticus / vulnificus, Hygiena™, Qualicon Diagnostics LLC
Catalog # 10754-892
Supplier:  Hygiena
Supplier Number:  KIT2010
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Sample Type:
    Tuna, Raw And Cooked Shrimp, Scallops, Oysters
  • Processing Time:
    70 minutes
  • Cat. No.:
    10754-892
  • Response time:
    18-20 hours
  • Target group:
    V. cholerae, V. parahaemolyticus, V. vulnificus
  • Supplier no.:
    D12863877
  • No. of tests:
    96

Specifications

About this item

The Hygiena™ BAX® System PCR Assay for Vibrio cholerae, parahaemolyticus, and vulnificusproduces clear yes-or-no results after a fast processing time in the BAX® System Q7 instrument.

The Hygiena BAX System Real-Time PCR Assay for Vibrio cholerae, parahaemolyticus, and vulnificus produces clear yes-or-no results for three major Vibrio strains in less than 24 hours after just 70 minutes of processing in the BAX System Q7 instrument. Certified as an AOAC-RI Performance Tested MethodSM for presence/absence or MPN testing for tuna, raw and cooked shrimp, scallops, and oysters.

The Hygiena BAX System is an automated detection system that uses the power of the polymerase chain reaction (PCR) to detect harmful bacteria and other microbes in food ingredients, finished products and environmental surfaces.

Ordering information: Includes PCR tubes with tablets, optical caps, protease, and lysis buffer. This assay can only be used with the BAX System Q7 instrument.