About this item
The Hygiena™ BAX® System PCR Assay for Listeria
L. monocytogenes produces clear yes-or-no results after a fast processing time in the BAX® System Q7 instrument. The Hygiena BAX System Real-Time PCR Assay for L. monocytogenes produces clear yes-or-no results after about 70 minutes of processing in the BAX System Q7 instrument. This assay is compatible with the BAX System Real-Time Salmonella and Real-Time Genus Listeria assays to detect all three targets in a single batch. The Hygiena BAX System Real-Time PCR Assay for Genus Listeria produces clear yes-or-no results for Listeria species after about 70 minutes of processing in the BAX System Q7 instrument. Certified as an AOAC-RI Performance Tested MethodSM for dairy, ready-to-eat meat, seafood, vegetable and environmental samples. TheHygiena BAX System PCR Assay for L. monocytogenes produces clear yes-or-no results after about 3.5 hours of processing in the BAX System Q7 instrument. Certified as a Performance Tested MethodSM and an Official Method of Analysis by AOAC on a wide variety of food and environmental sample types. Adopted by the USDA-FSIS for processed meat, poultry, pasteurized liquid egg products and environmental sponge samples.
Offering single-step enrichment and delivering next-day results, the DuPont BAX System PCR Assay for L. monocytogenes 24E produces clear yes-or-no results after about 3.5 hours of processing in the BAX System Q7 instrument. Certified as an AOAC-RI Performance Tested MethodSM for meat, seafood, dairy, and environmental samples. Validated by AFNOR Certification for all human food products and environmental samples. The Hygiena BAX System PCR Assay for Genus Listeria produces clear yes-or-no results for Listeria species after about 3.5 hours of processing in the BAX System Q7 instrument. Certified as a Performance Tested MethodSM for frankfurters, smoked salmon, vegetables, cheese and environmental samples.
Offering single-step enrichment and delivering next-day results, the DuPont BAX System PCR Assay for Genus Listeria 24E produces clear yes-or-no results for Listeria spp. after about 3.5 hours of processing in the BAX System Q7 instrument. Certified as a Performance Tested MethodSM for meat, seafood, dairy, and environmental samples. The Hygiena BAX System is an automated detection system that uses the power of the polymerase chain reaction (PCR) to detect harmful bacteria and other microbes in food ingredients, finished products and environmental surfaces.
Specifications
- Sample Type:Raw And Processed Meats, Seafood, Dairy Cultured/Non-Cultured, Eggs And Egg Products, Fresh Produce/Vegetables, Fruit Juices
- Processing Time:3.5 hours