About this item
The Hygiena™ BAX® System PCR Assay for Salmonella produces clear yes-or-no results after a fast processing time in the BAX® System Q7 instrument.
Delivering results in as few as 12 hours for some matrices, the DuPont BAX System Real-Time PCR Assay for Salmonella produces clear yes-or-no results for Salmonella species after about 70 minutes of processing in the BAX System Q7 instrument. Certified as a Performance Tested MethodSM and an Official Method of Analysis by AOAC on a wide variety of food and environmental sample types.
Delivering next-day results, the DuPont BAX System PCR Assay for Salmonella produces clear yes-or-no results for Salmonella species after about 3.5 hours of processing in the BAX System Q7 instrument. Certified as a Performance Tested MethodSM and an Official Method of Analysis by AOAC on a wide variety of food and environmental sample types. Validated by AFNOR Certification for all human food products and environmental samples. Adopted by the USDA-FSIS for testing meat, poultry, pasteurized egg, and catfish products and carcass and environmental sponges.
Using proprietary “hot-start” technology, the second-generation DuPont BAX System PCR Assay for Salmonella 2 keeps the reaction enzyme inactive until PCR begins. This assay produces clear yes-or-no results for Salmonella species after about 3.5 hours of processing in the BAX System Q7 instrument. Certified as a Performance Tested MethodSM and an Official Method of Analysis by AOAC on a wide variety of food and environmental sample types. Validated by AFNOR Certification for all human food products and environmental samples.
The Hygiena BAX System is an automated detection system that uses the power of the polymerase chain reaction (PCR) to detect harmful bacteria and other microbes in food ingredients, finished products and environmental surfaces.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Sample Type:Ground Beef, Trim, Produce Dairy, Chocolate/Bakery Products, Pasta Dry Pet Food, Environmental Samples
- Processing Time:3.5 hours