Anti-ZASP/ CYPHER / LDB3 (LIM Domain binding 3) Goat Polyclonal Antibody
89297-676
: Genetex
- Antibody Type:Primary
- Antigen Name:Lim Domain Binding 3
- Antigen Symbol:ZASP/ CYPHER / LDB3 (LIM Domain binding 3)
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Pig,Human,Dog,Rat,Mouse
- Western Blot:Yes
- Size:100 μg
- Gene ID:11155
- Antigen Synonyms:ZASP|CMD1C|PDLIM6|LDB3Z4|LVNC3|MFM4|LDB3Z1|ORACLE|CMH24|CYPHER|CMPD3
- Storage Buffer:Tris saline, 0.02% sodium azide, pH7.3 with 0.5% bovine serum albumin.
- Storage Temperature:Aliquot and store at –20 °C. Minimize freezing and thawing.
- Concentration:0.5mg/ml
- Immunogen:C-KSQNKPEDEADE
- Purification:Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
- Cat. No.:89297-676
- Supplier no.:GTX89478
Goat polyclonal antibody to ZASP/ CYPHER / LDB3
Experiments gave bands at approx 80-90kDa and 30-35kDa in Human Skeletal Muscle lysates after 0.01µg/ml antibody staining. These bands correspond to earlier findings in literature with different antibodies (Huang et al J Biol Chem. 2003 Feb 28;278(9):7360-5; PMID: 12499364). This protein has a calculated MW of 77.1kDa according to NP_009009.1. Recommended concentration: 0.01-0.03µg/ml.
Type: Primary
Antigen: ZASP/ CYPHER / LDB3 (LIM Domain binding 3)
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse, Dog, Pig, Rat